Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega TNT™ T7 Insect Cell Extract Protein Expression System Companion Vectors
Click to view available options
Vector:
pF25A ICE T7 Flexi vector
pF25K ICE T7 Flexi vector
Description
- Protein Expression Vectors Used With Insect Cell Extracts for In Vitro Translation
- Efficient transfer of Flexi(R) vector inserts for direct use of recombinant clones, minimizing time wasted screening background colonies
- Adaptable to high-throughput formats for large screening projects
- Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or mammalian protein expression) as needed
Specifications
Specifications
Promoter | T7 |
Restriction Site | SgfI and PmeI |
Storage Buffer | 10mM Tris HCl, 1mM EDTA (pH 8) |
Content And Storage | -30°C to -10°C |
Concentration | 100 ng/μL |
Quantity | 20 μg |
Vector | pF25A ICE T7 Flexi vector |
For Use With (Application) | Clone ORF and conveniently transfer these inserts to a variety of vectors designed for multiple applications |
- Patent Pending.
- For research use only. Persons wishing to use this product or its derivatives in other fields of use, including without limitation, commercial sale, diagnostics or therapeutics, should contact Promega Corporation for licensing information.
- Use of this vector for applications outside of the TNT T7 Insect Cell Extract Protein Expression System may require a license from Shimadzu Corporation. For more information contact: t-direct@shimadzu-biotech.jp.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction