Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega TRKA Kinase Enzyme System

Catalog No. PRV2931 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2931 10 μg
PRV9761 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV2931 Supplier Promega Supplier No. V2931
Add to Cart
Add to Cart

Recombinant human TRKA (amino acids 440-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TRKA is a member of the trk proto-oncogene family and encodes a 140kDa membrane-spanning protein tyrosine kinase.

  • Easily Screen and Profile TRKA Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Decreased expression of TRKA on striatal cholinergic neurons may contribute to alzheimer disease
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.