Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega UGT-Glo™ UGT1A1 Screening System
Click to view available options
Quantity:
1000 Assays
Description
- Speed: The luminescent format eliminates the need for time-consuming analyses such as HPLC and LC/MS.
- Simplified Method: The simple“add and read”protocol makes the assay amenable to higher throughput screening in multiwell plates.
- Sensitive: Allows researchers to use less enzyme and scale down reaction volumes, which saves on reagent costs.
Specifications
Specifications
Includes | 50mM UDPGA, 5X UGT Buffer, UGT multienzyme Substrate, UGT2B7 microsomes, 100X D-cysteine, UGT1A4 Substrate, luciferin detection reagent, Reconstitution Buffer, control membranes |
Quantity | 1000 Assays |
Content And Storage | -70°C |
For Use With (Application) | Provides a luminescent method for measuring UDP Glucuronosyltransferase activity |
Product Type | UGT-Glo UGT2B7 screening system |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction