Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ViviRen™ Live Cell Substrate

Catalog No. PRE6492 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
0.37 mg
3.7 mg
37 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE6492 3.7 mg
PRE6491 0.37 mg
PRE6495 37 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRE6492 Supplier Promega Supplier No. E6492
Add to Cart
Add to Cart

ViviRen Live Cell Substrate is a uniquely engineered coelenterazine that generates three- to fivefold brighter Renilla luciferase luminescence for super-sensitive quantitation of reporter gene, BRET and RNAi activity.

  • Sensitive Quantitation of Renilla Luminescence in Live Cells
  • Engineered coelenterazine for live-cell reporter gene, BRET and RNAi assays
  • Permits real-time measurement in multiwell plates
  • High signal-to-background ratios

  • U.S. Pat. Nos. 7,268,229 and 7,537,912, Canadian Pat. No. 2,462,506 and other patents and patents pending.
  • Certain applications of this product may require licenses from others.
  • This product does not convey a license to use recombinant Renilla luciferase under U.S. Patent 5,418,155 and related patents. Promega offers licensed Renilla luciferase vectors, which may be used in conjunction with this product.

Specifications

Product Type Live Cell Substrate
Content And Storage <-10°C
Quantity 3.7 mg
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.