Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ MagneSil™ Plasmid Purification System

Catalog No. PRA1631 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
100 x 96 Preps
4 x 96 Preps
8 x 96 Preps
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA1631 8 x 96 Preps
PRA1630 4 x 96 Preps
PRA1641 100 mL
PRA1635 100 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRA1631 Supplier Promega Supplier No. A1631
Add to Cart
Add to Cart

Includes

  • MagneSil BLUE, MagneSil RED, plus necessary solutions, buffers, and collection plates for plasmid purification
  • MagneSil RED (also available separately)

Requires

  • Orbital microplate shaker
  • Magnetic workstation that will accommodate a microplate, such as the MagnaBot™ 96 Magnetic Separation Device (PR-V8151, sold separately)

The Wizard™ MagneSil Plasmid DNA Purification System provides a simple, reliable method for rapid isolation of plasmid DNA in a 96-well, high-throughput format.

  • Rapid Isolation of Plasmid DNA in 96-Well Format
  • Process multiple plates without user intervention
  • Validated automated methods available

  • U.S. Patents 6,284,470 and 7,078,224; Australian Patent 778486 and other patents pending.
  • U.S. Patents 6,027,945 and 6,368,800; Australian Patent 732756 and other patents and patents pending.

Specifications

Content And Storage 15°C to 30°C
Description Wizard MagneSil Plasmid Purification System
Format 96-well Plate
For Use With (Equipment) Wizard MagneSil Plasmid Purification System
For Use With (Application) Simple and reliable method for the rapid isolation of plasmid DNA
Includes
  • MagneSil BLUE, MagneSil RED, plus necessary solutions, buffers, and collection plates for plasmid purification MagneSil RED (also available separately)
Product Type Plasmid Purification
Quantity 8 x 96 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.