Learn More
Description
Includes
- MagneSil BLUE, MagneSil RED, plus necessary solutions, buffers, and collection plates for plasmid purification
- MagneSil RED (also available separately)
Requires
- Orbital microplate shaker
- Magnetic workstation that will accommodate a microplate, such as the MagnaBot™ 96 Magnetic Separation Device (PR-V8151, sold separately)
- Rapid Isolation of Plasmid DNA in 96-Well Format
- Process multiple plates without user intervention
- Validated automated methods available
- U.S. Patents 6,284,470 and 7,078,224; Australian Patent 778486 and other patents pending.
- U.S. Patents 6,027,945 and 6,368,800; Australian Patent 732756 and other patents and patents pending.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | 96-well Plate |
| For Use With (Equipment) | Wizard MagneSil Plasmid Purification System HTP1 |
| For Use With (Application) | Simple and reliable method for the rapid isolation of plasmid DNA |
| Includes | 3 x 500mL Cell resuspension solution, 3 x 500mL Cell Lysis solution, 3 x 500mL neutralization solution, 7 x 100mL Magnesil™ blue, 7 x 100mL Magnesil™ red, 3 x 500mL Elution Buffer, 100 Collection plates, protocol |
| Product Type | Plasmid Purification |
| Quantity | 100 x 96 Preps |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.