Learn More
Promega Wizard™ PureFection Plasmid DNA Purification System1, 2
Click to view available options
Quantity:
10 midipreps/2 maxipreps
125 midipreps/25 maxipreps
25 midipreps/5 maxipreps
Description
- Utilizes MagneSil Paramagnetic Particles, a magnetic separation technology that allows rapid purification
- Suitable for both midipreps (5 to 25mL of bacterial culture) and maxipreps (26 to 125mL of bacterial culture)
- Produces DNA suitable for use in a variety of molecular biology applications, including transfection
- Does not require phenol extractions or precipitations
- Store at 22° to 25°C
Note: PR-A2160 and PR-A2170 do not contain the magnetic separation unit (PR-A2231); this unit must be purchased separately
- U.S. Patents 6,027,945; 6,368,800; and 6,673,631; Australian Patent 732756 and other patents and patents pending.
- U.S. Patent 6,194,562; Australian Patent 740145; Canadian Patent 2,329,067 and other patents pending.
Specifications
Specifications
Quantity | 125 midipreps/25 maxipreps |