Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ SV 96 Lysate Clearing and Binding Plates

Catalog No. PRA2241 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
SV96 Binding Plate
SV96 Lysate Clearning Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PR-A2241 SV96 Lysate Clearning Plate
PR-A2271 SV96 Binding Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-A2241 Supplier Promega Supplier No. A2241
Add to Cart
Add to Cart

For use with Wizard SV 96 and Wizard SV 9600 Plasmid DNA Purification Systems

  • Use with several Wizard(R) DNA purification systems or the SV 96 Total RNA Isolation System
  • Perform isolation manually or on a robotic platform
  • Use with the Vac-Man(R) 96 Vacuum Manifold

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Nucleic acid purification
Product Type SV96 Lysate Clearning Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.