Learn More
Promega Wizard SV Gel and PCR Clean-Up System and x-tracta™ Gel Extractor Package
Catalog No. PRA9284 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 Preparation/100 Extractors
50 Preparation/25 Extractors
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA9284 250 Preparation/100 Extractors
PRA9283 50 Preparation/25 Extractors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA9284 Supplier Promega Supplier No. A9284
Add to Cart
Add to Cart
The Wizard™ SV Gel and PCR Clean-Up System is designed to purify DNA fragments of 100bp-10kb from standard or low-melting agarose gels or to purify products directly from PCR or restriction digests.
  • Purifies DNA Fragments from Gels or PCR
  • Purifies 100bp to 10kb DNA fragments
  • Includes gel extractors for added speed and convenience
Specifications
Type Wizard™ SV Gel and PCR Clean up System and x-tracta™ Gel Extractor Package
Quantity 250 Preparation/100 Extractors
Temperature (Metric) Storage 22°C to 25°C