Learn More
Promega x-tracta™ Gel Extractors
Catalog No. PRA2121 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100/Pk.
25/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA2121 25/Pk.
PRA2122 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA2121 Supplier Promega Supplier No. A2121
Add to Cart
Add to Cart
The x-tracta Gel Extractor tool provides a convenient, safe method for removal of agarose gel fragments for further processing. The device removes a 0.13 x 0.33 inch gel piece from agarose gels for easy transfer into a microcentrifuge tube for processing.
  • Safe, Convenient Extraction of Gel Slices
  • Removes a 0.13 x 0.33 inch gel piece
  • Disposable device reduces cross-contamination
  • No need to use razor blades or scalpels
Specifications
Type x-tracta™ Gel Extractor
Quantity 25/Pk.
Temperature (Metric) Storage 15°C to 30°C