Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prominin 2 Antibody (244029), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2024L
Description
Prominin 2 Monoclonal antibody specifically detects Prominin 2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
Prominin 2 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
150696 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunocytochemistry, CyTOF | |
244029 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
hPROML2, MGC138714, PROM-2, prominin 2, prominin-2, Prominin-like protein 2, prominin-related protein, PROML2 | |
NS0 mouse myeloma cell line transfected with human Prominin 2, Ala27-Leu834, Accession # AAM10541 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction