Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prominin 2 Antibody (244029), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2024MFV450
Description
Prominin 2 Monoclonal antibody specifically detects Prominin 2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| Prominin 2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 244029 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| hPROML2, MGC138714, PROM-2, prominin 2, prominin-2, Prominin-like protein 2, prominin-related protein, PROML2 | |
| NS0 mouse myeloma cell line transfected with human Prominin 2, Ala27-Leu834, Accession # AAM10541 | |
| RUO | |
| 150696 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction