Learn More
Invitrogen™ Prominin 2 Monoclonal Antibody (244029)

Description
This antibody stains human Prominin 2-transfected cells but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Prominin 2 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 244029 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PROM2 |
| Gene Accession No. | Q8N271 |
| Gene Alias | AI507243; h prominin-2; hPROML2; mPROML2; PROM2; PROM-2; prominin 2; prominin-2; Prominin-like protein 2; prominin-related protein; PROML2; Promrp; Prom-rp; rPROML2; Testosterone-regulated prominin-related protein; Trprp; UNQ2521/PRO6014 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.