Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prominin 2 Monoclonal Antibody (OTI5E5)
GREENER_CHOICE

Catalog No. PIMA524902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524902 Supplier Invitrogen™ Supplier No. MA524902
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prominin 2 Monoclonal Antibody for Western Blot, ICC/IF, IP

This gene encodes a member of the prominin family of pentaspan membrane glycoproteins. The encoded protein localizes to basal epithelial cells and may be involved in the organization of plasma membrane microdomains. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prominin 2
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PROM2
Gene Accession No. Q8N271
Gene Alias AI507243; h prominin-2; hPROML2; mPROML2; PROM2; PROM-2; prominin 2; prominin-2; Prominin-like protein 2; prominin-related protein; PROML2; Promrp; Prom-rp; rPROML2; Testosterone-regulated prominin-related protein; Trprp; UNQ2521/PRO6014
Gene Symbols PROM2
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human PROM2 expression vector.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150696
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.