Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prominin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584613
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584613 Supplier Invitrogen™ Supplier No. PA584613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELGADFSQVP SVDHVLHQLK GVPEANFSSM VQEENSTFNA LPALAAMQTS SVVQELKKAV AQQPEGVRTL AEGFP Highest antigen sequence indentity to the following orthologs: Mouse - 67%, Rat - 73%.

This gene encodes a member of the prominin family of pentaspan membrane glycoproteins. The encoded protein localizes to basal epithelial cells and may be involved in the organization of plasma membrane microdomains. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prominin 2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PROM2
Gene Accession No. Q8N271
Gene Alias AI507243; h prominin-2; hPROML2; mPROML2; PROM2; PROM-2; prominin 2; prominin-2; Prominin-like protein 2; prominin-related protein; PROML2; Promrp; Prom-rp; rPROML2; Testosterone-regulated prominin-related protein; Trprp; UNQ2521/PRO6014
Gene Symbols PROM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PROM2. Recombinant protein control fragment (Product #RP-103524).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.