Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prominin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532995
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532995 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532995 Supplier Invitrogen™ Supplier No. PA532995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset (100%) Bovine, Horse (94%) Gibbon, Bat, Rabbit (88%) Hamster, Elephant (81%).

This gene encodes a member of the prominin family of pentaspan membrane glycoproteins. The encoded protein localizes to basal epithelial cells and may be involved in the organization of plasma membrane microdomains. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prominin 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PROM2
Gene Accession No. Q8N271
Gene Alias AI507243; h prominin-2; hPROML2; mPROML2; PROM2; PROM-2; prominin 2; prominin-2; Prominin-like protein 2; prominin-related protein; PROML2; Promrp; Prom-rp; rPROML2; Testosterone-regulated prominin-related protein; Trprp; UNQ2521/PRO6014
Gene Symbols PROM2
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminus of human PROM2.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150696
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.