Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PROP1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 1 publication
$487.50
Specifications
Antigen | PROP1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Peptide ELISA Detection limit 1:32000 |
Applications | ELISA |
Classification | Polyclonal |
Description
PROP1 Polyclonal specifically detects PROP1 in Human samples. It is validated for Peptide ELISA.Specifications
PROP1 | |
Peptide ELISA Detection limit 1:32000 | |
Polyclonal | |
Goat | |
Human | |
NP_006252.2 | |
5626 | |
Peptide with sequence AERRRQAEKPKKGR-C corresponding to N-Terminus according to NP_006252.2. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
ELISA | |
Unconjugated | |
RUO | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CPHD2, homeobox protein prophet of Pit-1, Pituitary-specific homeodomain factor, PROP paired-like homeobox 1, PROP-1, prophet of Pit1, paired-like homeodomain transcription factor | |
PROP1 | |
IgG | |
Affinity Purified | |
This antibody is specific to the N-Terminus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title