Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116708 Supplier Invitrogen™ Supplier No. PA5116708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain Immunogen sequence: GGGRSRFSPQ GGQRGRPHSR RRHRTTFSPV QLEQLESAFG RNQYPDIWAR ESLARDTGLS EARIQVWFQN RRAKQRKQER SLLQPLAHLS PAAFSSFLPE S.

PROP1 encodes a paired-like homeodomain transcription factor in the developing pituitary gland. Expression occurs prior to and is required for expression of pou domain transcription factor 1, which is responsible for pituitary development and hormone expression. Mutations in this gene have been associated with combined pituitary hormone deficiency-2 as well as deficiencies in luteinizing hormone, follicle-stimulating hormone, growth hormone, prolactin, and thyroid-stimulating hormone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PROP1
Gene Accession No. P97458
Gene Alias Ames dwarf; CPHD2; df; Homeobox protein prophet of Pit-1; paired like homeodomain factor 1; pituitary specific homeodomain factor; pituitary-specific homeodomain factor; PROP paired-like homeobox 1; PROP1; PROP-1; prophet of pit-1; prophet of Pit1 paired-like homeodomain transcription factor; prophet of Pit1, paired-like homeodomain transcription factor; prophet of POU1F1; transcription factor
Gene Symbols PROP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human PROP1 (NP_0062523).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19127
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.