Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100890 Supplier Invitrogen™ Supplier No. PA5100890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PROP1.

PROP1 encodes a paired-like homeodomain transcription factor in the developing pituitary gland. Expression occurs prior to and is required for expression of pou domain transcription factor 1, which is responsible for pituitary development and hormone expression. Mutations in this gene have been associated with combined pituitary hormone deficiency-2 as well as deficiencies in luteinizing hormone, follicle-stimulating hormone, growth hormone, prolactin, and thyroid-stimulating hormone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PROP1
Gene Accession No. O75360, P97458
Gene Alias Ames dwarf; CPHD2; df; Homeobox protein prophet of Pit-1; paired like homeodomain factor 1; pituitary specific homeodomain factor; pituitary-specific homeodomain factor; PROP paired-like homeobox 1; PROP1; PROP-1; prophet of pit-1; prophet of Pit1 paired-like homeodomain transcription factor; prophet of Pit1, paired-like homeodomain transcription factor; prophet of POU1F1; transcription factor
Gene Symbols PROP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PROP1(Accession O75360), corresponding to amino acid residues A98-P148.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19127, 266738, 5626
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.