Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ProPac™ 3R SCX 3 μm HPLC Columns

Catalog No. 43103052068
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
Diameter (Metric):
2 mm
4 mm
Flow Rate:
0.1 to 0.2 mL/min.
0.1 to 0.3 mL/min
0.3 to 0.5 mL/min
0.3 to 0.5 mL/min.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Flow Rate
43-103-052068 50 mm 2 mm 0.1 to 0.3 mL/min
43-103-054068 50 mm 4 mm 0.3 to 0.5 mL/min
43-103-102068 100 mm 2 mm 0.1 to 0.2 mL/min.
43-103-104068 100 mm 4 mm 0.3 to 0.5 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 43-103-052068 Supplier Thermo Scientific™ Supplier No. 43103052068
Add to Cart
Add to Cart
This item is not returnable. View return policy

Get the performance you need with ProPac™ 3R SCX HPLC columns. Suitable for charge variant separations and recommended for therapeutic proteins, mAbs, ADCs, and biosimilar proteins.

Use Thermo Scientific™ ProPac™ 3R SCX 3 μm HPLC columns to provide the reproducibility, resolution, and robustness needed for protein characterization. These columns are ideal for charge variant analysis and recommended for use with therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), and biosimilar proteins.

Designed with bio-inert materials to reduce secondary interactions, our ProPac 3R SCX columns deliver the performance you need over a broad range of pH's, temperatures, and mobile phase compositions with excellent recovery and low carryover.

Suitable for use with mass spectrometry (MS).

Maximizing Reproducibility, Resolution, and Robustness

The ProPac 3R SCX 3 μm HPLC column is a strong cation-exchanger with a quaternary amine functional group, suitable for use in biomolecule applications. These specialized ion-exchange chromatography columns provide qualitative and quantitative evaluation of charge variant analysis to biopharmaceutical scientists who need reproducible resolving capabilities when identifying and quantifying charge variant profiles of proteins.

Our ProPac 3R SCX columns offer superior performance for characterization of proteins including monoclonal antibodies (mAbs), therapeutic proteins, antibody drug conjugates (ADCs), and biosimilar proteins.

Column Features:

  • Ideal for characterization and quality control assessment of closely-related protein variants
  • Unequaled high-resolution separations
  • High-efficiency peaks with excellent recovery
  • Higher capacity than other columns based on nonporous particles
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Compatible with mass spectrometry (MS)

We recommend our CX-1 pH gradient buffers when using these columns with LC-UV/LC-FLD.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2 mm
Flow Rate 0.1 to 0.3 mL/min
Length (Metric) 50 mm
Max. Pressure 4500 psi (310 bar)
Mobile Phase Compatibility Recommended for use with CX-1 pH buffers
Particle Size 3 μm
pH 2 to 12
Pore Size Nonporous
Temperature Ambient to 60°C
Stationary Phase Proprietary
Product Line ProPac
Packing Material Monodisperse Polymer Resin
Column Type Ion Exchange
Phase Strong Cation Exchange
Material PEEK Hardware
Technique Bio-LC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.