Learn More
Description
Use Thermo Scientific™ ProPac™ 3R SCX 3 μm HPLC columns to provide the reproducibility, resolution, and robustness needed for protein characterization. These columns are ideal for charge variant analysis and recommended for use with therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs), and biosimilar proteins.
Designed with bio-inert materials to reduce secondary interactions, our ProPac 3R SCX columns deliver the performance you need over a broad range of pH's, temperatures, and mobile phase compositions with excellent recovery and low carryover.
Suitable for use with mass spectrometry (MS).
Maximizing Reproducibility, Resolution, and Robustness
The ProPac 3R SCX 3 μm HPLC column is a strong cation-exchanger with a quaternary amine functional group, suitable for use in biomolecule applications. These specialized ion-exchange chromatography columns provide qualitative and quantitative evaluation of charge variant analysis to biopharmaceutical scientists who need reproducible resolving capabilities when identifying and quantifying charge variant profiles of proteins.
Our ProPac 3R SCX columns offer superior performance for characterization of proteins including monoclonal antibodies (mAbs), therapeutic proteins, antibody drug conjugates (ADCs), and biosimilar proteins.
Column Features:
- Ideal for characterization and quality control assessment of closely-related protein variants
- Unequaled high-resolution separations
- High-efficiency peaks with excellent recovery
- Higher capacity than other columns based on nonporous particles
- Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
- Compatible with mass spectrometry (MS)
We recommend our CX-1 pH gradient buffers when using these columns with LC-UV/LC-FLD.
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4 mm |
| Flow Rate | 0.3 to 0.5 mL/min |
| Length (Metric) | 50 mm |
| Max. Pressure | 4500 psi (310 bar) |
| Mobile Phase Compatibility | Recommended for use with CX-1 pH buffers |
| Particle Size | 3 μm |
| pH | 2 to 12 |
| Pore Size | Nonporous |
| Temperature | Ambient to 60°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.