Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl alcohol, 99%

Catalog No. AAA10295AP
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 107-19-7 | C3H4O | 56.06 g/mol

Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

Solubility
Fully miscible in water.

Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-19-7
Molecular Formula C3H4O
Molecular Weight (g/mol) 56.06
MDL Number MFCD00002912
InChI Key TVDSBUOJIPERQY-UHFFFAOYSA-N
Synonym propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
PubChem CID 7859
ChEBI CHEBI:28905
IUPAC Name prop-2-yn-1-ol
SMILES OCC#C

Specifications

Melting Point -53°C
Density 0.949
Boiling Point 114°C to 115°C
Flash Point 33°C (91°F)
Refractive Index 1.432
Linear Formula HC≡CCH2OH
Quantity 500 mL
UN Number UN2929
Beilstein 506003
Merck Index 14,7809
Solubility Information Fully miscible in water.
Formula Weight 56.06
Percent Purity 99%
Chemical Name or Material Propargyl alcohol
Show More Show Less
Hazard Category H226-H301-H310+H330-H314-H335-H373
Hazard Statement GHS H Statement
H301-H311-H331-H314-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 203-471-2
RTECSNumber UK5075000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.