Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
500mL
1L
2.5L
Packaging:
Glass bottle
107-19-7,50-00-0
C3H4O
56.06
MFCD00002912
TVDSBUOJIPERQY-UHFFFAOYSA-N
propargyl alcohol,2-propyn-1-ol,ethynylcarbinol,1-propyn-3-ol,propynyl alcohol,2-propynyl alcohol,2-propynol,3-propynol,1-hydroxy-2-propyne,3-hydroxy-1-propyne
7859
CHEBI:28905
prop-2-yn-1-ol
C#CCO
107-19-7,50-00-0
C3H4O
56.06
MFCD00002912
TVDSBUOJIPERQY-UHFFFAOYSA-N
propargyl alcohol,2-propyn-1-ol,ethynylcarbinol,1-propyn-3-ol,propynyl alcohol,2-propynyl alcohol,2-propynol,3-propynol,1-hydroxy-2-propyne,3-hydroxy-1-propyne
7859
CHEBI:28905
prop-2-yn-1-ol
C#CCO
Density 0.9490g/mL
Viscosity 1.58 mPa.s (20°C)
Infrared Spectrum Authentic
Color Colorless to Yellow
Specific Gravity 0.949
Flash Point 34°C
Boiling Point 114.0°C to 115.0°C
Packaging Glass bottle
Melting Point -53.0°C
Quantity 500mL
Refractive Index 1.4300 to 1.4330
Assay Percent Range 99%
Linear Formula HC≡CCH2OH
Beilstein 01,454
Fieser 09,394
Merck Index 15,7923
Solubility Information Solubility in water: miscible. Other solubilities: miscible with benzene, chloroform, ethanol, ether,, 1,2-dichloroethane, acetone, dioxane, thf,pyridine, moderately soluble in ccl4, immiscible with aliphatic hydrocarbons
Formula Weight 56.06
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Propargyl alcohol, 99%
Show More Show Less

GHS P Statement
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Fatal if inhaled.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

;GHS P StatementWear protective gloves/protective clothing/eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing.Immediately call a POISO

EINECSNumber : 203-471-2

RTECSNumber : UK5075000

TSCA : TSCA