Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propargyl chloride, 70 wt.% solution in toluene
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Density | 0.9630g/mL |
Boiling Point | 60.0°C to 63.0°C |
Flash Point | −15°C |
Assay Percent Range | 68 to 72% |
Packaging | Glass bottle |
Linear Formula | HC≡CCH2Cl |
Quantity | 100 mL |
Beilstein | 01,248 |
Fieser | 05,565 |
Merck Index | 15,7924 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Suspected of damaging the unborn child. May cause dro |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call |
EINECSNumber | 210-856-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction