Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. P23361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23361G 1 g
P23365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23361G Supplier TCI America Supplier No. P23361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13702-09-5
Molecular Formula C6H6S
Molecular Weight (g/mol) 110.174
MDL Number MFCD00048109
InChI Key XPTMJJIPRSWBDK-UHFFFAOYSA-N
Synonym Dipropargyl Sulfide
PubChem CID 518812
IUPAC Name 3-prop-2-ynylsulfanylprop-1-yne
SMILES C#CCSCC#C

Specifications

Boiling Point 59°C
Quantity 1 g
UN Number 1993
Formula Weight 110.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propargyl Sulfide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.