Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargylamine, 98%

Catalog No. aah5349506
Click to view available options
Quantity:
5 g
25 g

CAS: 2450-71-7 | C3H5N | 55.08 g/mol

Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.

Solubility
Miscible with water, chloroform and ethyl acetate.

Notes
Hygroscopic. Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2450-71-7
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00008198
InChI Key JKANAVGODYYCQF-UHFFFAOYSA-N
Synonym propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine
PubChem CID 239041
IUPAC Name prop-2-yn-1-amine
SMILES C#CCN

Specifications

Density 0.86
Boiling Point 83°C
Flash Point 6°C (43°F)
Refractive Index 1.449
Linear Formula HC≡CCH2NH2
Quantity 5 g
UN Number UN3286
Beilstein 773681
Sensitivity Air Sensitive
Solubility Information Miscible with water,chloroform and ethyl acetate.
Formula Weight 55.08
Percent Purity 98%
Chemical Name or Material Propargylamine
Show More Show Less
Hazard Category H225-H302-H311-H314
Hazard Statement GHS H Statement
H225-H310-H314-H318-H302
Highly flammable liquid and vapor.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber 219-513-8
RTECSNumber UK5250000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.