Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Properdin Monoclonal Antibody (KT20)

Catalog No. MA172515
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA172515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA172515 Supplier Invitrogen™ Supplier No. MA172515

Mouse Monoclonal Antibody

This antibody can be used as a primary antibody for ELISA(1 μg/mL).

This gene encodes a plasma glycoprotein that positively regulates the alternative complement pathway of the innate immune system. This protein binds to many microbial surfaces and apoptotic cells and stabilizes the C3- and C5-convertase enzyme complexes in a feedback loop that ultimately leads to formation of the membrane attack complex and lysis of the target cell. Mutations in this gene result in two forms of properdin deficiency, which results in high susceptibility to meningococcal infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Properdin
Applications ELISA
Classification Monoclonal
Clone KT20
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CFP
Gene Accession No. P27918
Gene Alias BCFG; Bf; BFD; C3 Proaccelerator; C3/C5 convertase; cfb; CFP; Complement factor P; complement factor properdin; GBG; H2-Bf; PBF2; PFC; PFD; PROPERDIN; properdin factor, complement; properdin P factor, complement
Gene Symbols CFP
Host Species Mouse
Immunogen Human properdin purified from blood.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5199
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.