Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Properdin Mouse anti-Human, Unconjugated, Clone: 6E9E6, MilliporeSigma™

Catalog No. MABF2125100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2125-100 100 μL
MABF2125-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2125-100 Supplier MilliporeSigma Supplier No. MABF2125100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Properdin, clone 6E9E6 Antibody, Cat. No. MABF2125, is a mouse monoclonal antibody that detects Properdin and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Properdin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6E9E6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias Complement factor P
Gene Symbols CEP;PFC
Host Species Mouse
Immunogen Purified properdin from human plasma.
Molecular Weight of Antigen ∽51 kDa observed; 51.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001138724
Test Specificity Clone 6E9E6 specifically detects human Properdin.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.