Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Properdin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109520 Supplier Invitrogen™ Supplier No. PA5109520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPTHGAWATW GPWTPCSASC HGGPHEPKET RSRKCSAPEP SQKPPGKPCP GLAYEQRRCT GLPPCPVAGG WGPWGPVSPC PVTCGLGQTM EQRTCNHPVP QHGGPFCAGD ATRTHICNTA VPCPVDGEWD SWGEWSPCIR RNMKSISCQE IPGQQSRGRT CRGRKFDGHR CAGQQQDIRH CYSIQHCPLK GSWSEWSTWG LCMPPCGPNP TRARQRLCTP LLPKYPPTVS MVEGQGEKNV TFWGRPLPRC EELQGQKLVV EEKRPCLHVP ACKDPEEEEL.

This gene encodes a plasma glycoprotein that positively regulates the alternative complement pathway of the innate immune system. This protein binds to many microbial surfaces and apoptotic cells and stabilizes the C3- and C5-convertase enzyme complexes in a feedback loop that ultimately leads to formation of the membrane attack complex and lysis of the target cell. Mutations in this gene result in two forms of properdin deficiency, which results in high susceptibility to meningococcal infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Properdin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CFP
Gene Accession No. P11680, P27918
Gene Alias BCFG; Bf; BFD; C3 Proaccelerator; C3/C5 convertase; cfb; CFP; Complement factor P; complement factor properdin; GBG; H2-Bf; PBF2; PFC; PFD; PROPERDIN; properdin factor, complement; properdin P factor, complement
Gene Symbols CFP
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18636, 299314, 5199
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.