Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Properdin Polyclonal Antibody, Invitrogen™

Catalog No. PIPA516203
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA516203 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA516203 Supplier Thermo Scientific Supplier No. PA516203
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PA5-16203 targets Properdin in IF applications and shows reactivity with Human samples. The PA5-16203 immunogen is properdin from human serum.

This gene encodes a plasma glycoprotein that positively regulates the alternative complement pathway of the innate immune system. This protein binds to many microbial surfaces and apoptotic cells and stabilizes the C3- and C5-convertase enzyme complexes in a feedback loop that μgtimately leads to formation of the membrane attack complex and lysis of the target cell. Mutations in this gene result in two forms of properdin deficiency, which results in high susceptibility to meningococcal infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Properdin
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 1% carrier protein and 0.1% sodium azide; pH 7.4-7.8
Gene CFP
Gene Accession No. P27918
Gene Alias BCFG; Bf; BFD; C3 Proaccelerator; C3/C5 convertase; cfb; CFP; Complement factor P; complement factor properdin; GBG; H2-Bf; PBF2; PFC; PFD; PROPERDIN; properdin factor, complement; properdin P factor, complement
Gene Symbols CFP
Host Species Goat
Immunogen Properdin from human serum
Purification Method purified
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5199
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.