Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Propidium Iodide ReadyProbes™ Reagent

Catalog No. R37108
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37108 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37108 Supplier Invitrogen™ Supplier No. R37108
  • $374.00
    $306.68 / Each of 1

    Save $67.32

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propidium Iodide ReadyProbes™ Reagent

Propidium iodide is a commonly used dead-cell stain that emits red fluorescence when bound to DNA. Propidium iodide is also used as a counterstain in multicolor fluorescent techniques. With Propidium Iodide ReadyProbes™ Reagent, we provide a room temperature-stable solution of classic DNA stain that comes in a convenient-to-use dropper bottle. Just tip and drip two drops per ml to stain your cells.

Features of Propidium Iodide ReadyProbes™ Reagent:
  • Ready-to-use liquid propidium iodide formulation
  • Rapid staining of dead cells without wash steps
  • Ready-to-use liquid formulation in convenient dropper bottle—no need to dilute, weigh, or pipette
  • Stable at room temperature—keep handy at your work station or cell culture area

    Classic Dead Cell Stain
    Propidium iodide is a classic fluorescent dye used extensively for staining of dead cells (Figure 1). It exhibits a sufficiently large Stokes shift to allow simultaneous detection of nuclear DNA and FITC or Alexa Fluor™ 488-labeled antibodies.

    Packaged for Convenience
    Propidium Iodide ReadyProbes™ Reagent is supplied ready-to-use and can be stored right next to your flow cytometer or in your cell culture lab. A liquid formulation in an ultra-convenient dropper bottle (Figure 2) allows you to stain your cells whenever and wherever they are ready. Just add two drops per ml for optimally bright and distinct nuclear counter-staining.

    Multiple Applications
    Propidium iodide is membrane-impermeant and excluded from viable cells. It is therefore commonly used for identifying dead cells in a population. Its spectral properties, selectivity for dsDNA, and increased fluorescence when bound to DNA, make propidium iodide ideal for use with green (Alexa Fluor™ 488, FITC, GFP) fluorophores in multicolor experiments. Because of its high affinity for DNA, it is also frequently used for counting cells, measuring apoptosis, and sorting cells based on DNA content.

    Suggestions for Use
  • In most cases, 2 drops/1×106 cells of Propidium Iodide ReadyProbes™ Reagent and an incubation time of 15 to 30 minutes will produce bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
  • When bound to nucleic acids, the fluorescence excitation maximum for propidium iodide is 535 nm and the emission maximum is 617 nm. Propidium iodide can be excited with a xenon or mercury-arc lamp or with the 488 line of an argon-ion laser. Generally, propidium iodide fluorescence is detected in the FL2 channel of flow cytometers.
  • As a preferred red nuclear stain in dead cell analyses using flow cytometry, Propidium Iodide ReadyProbes™ Reagent is ideal for use with antibody-based applications.

    See other Molecular Probes™ imaging tools and reagents.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Cell Permeability Cell-impermeant
    Color Far-red
    Content And Storage 6 x 2.5 mL dropper bottles

    Store at ≤ 25°C
    Detection Method Fluorescence
    For Use With (Application) Viability Assay
    For Use With (Equipment) Attune Acoustic Focusing Cytometer, EVOS Cell Imaging System, Flow Cytometer
    Product Type Nucleic Acid Stain
    Dye Type propidium iodide
    Form Liquid
    Product Line ReadyProbes
    Quantity 1 kit
    Sub Cellular Localization Nucleus
    Excitation Wavelength 535 nm
    Emission Wavelength 617
    Show More Show Less
    What is the volume for one drop from the ReadyProbes dropper bottles?

    The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

    Can I use the ReadyProbes reagents for flow cytometry?

    This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.