Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Pyrrole-2-carboxylic acid, 97%
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 g
                            
                        
                            
                                5 g
                            
                        
                            
                                25 g
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Melting Point | 207.0°C to 208.0°C | 
| Color | Pink to White | 
| Density | 0.862g/mL | 
| Infrared Spectrum | Authentic | 
| Assay Percent Range | 97% | 
| Packaging | Glass bottle | 
| Refractive Index | 1.4410 to 1.4450 | 
| Quantity | 25 g | 
| Beilstein | 22, 22 | 
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Warning | 
| Hazard Statement | GHS H Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.  | 
                                
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for  | 
                                
| EINECSNumber | 211-221-9 | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction