Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionamidoxime, 97%

Catalog No. p-7033731
Click to view available options
Quantity:
250 mg
1 g

CAS: 29335-36-2 | C3H8N2O | 88.11 g/mol

Propionamidoxime is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionamidoxime is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29335-36-2
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD03426272
InChI Key RLZPCFQNZGINRP-UHFFFAOYSA-N
Synonym n-hydroxypropionamidine, n-hydroxypropionimidamide, 1z-n'-hydroxypropanimidamide, 1-hydroxyimino propylamine, n-hydroxy-propionamidine, n-hydroxyproponmdamde, n-hydroxypropanamidine, n'-hydroxypropimidamide, z-n'-hydroxypropimidamide, e-n'-hydroxypropanimidamide
PubChem CID 6521662
IUPAC Name N'-hydroxypropanimidamide
SMILES CCC(=NO)N

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 88.11
Percent Purity 97%
Chemical Name or Material Propionamidoxime
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.