Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel(II) nitrate hexahydrate, 99%, for analysis
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
For analysis
Specifications
Specifications
| Boiling Point | 137.0°C |
| Melting Point | 56.7°C |
| Color | Blue to Green |
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Physical Form | Crystalline Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 98.5% min. (Complexometry) |
| Linear Formula | Ni(NO3)2·6H2O |
| Merck Index | 15,6598 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause cancer by inhalation. Suspected of causing genetic defects. May damage the unborn child. Causes damage to organs through prolonged or repeated exposure. Harmful if inhaled. Harmful if swallow |
| Precautionary Statement | GHS P Statement Use personal protective equipment as required. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF exposed or concerned: Get medical advice/attention. Store locked up. Avoid release to the e |
| RTECSNumber | QR7300000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction