Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) nitrate hexahydrate, 99%, for analysis

Catalog No. AC223150100
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle

CAS: 13478-00-7 | H12N2NiO12 | 290.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-00-7, 13138-45-9
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
PubChem CID 61630
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 137.0°C
Melting Point 56.7°C
Color Blue to Green
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 10 kg
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Ni(NO3)2·6H2O
Merck Index 15,6598
Solubility Information Solubility in water: 940g/L (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
IUPAC Name nickel(2+);dinitrate;hexahydrate
Formula Weight 290.8
Percent Purity 99%
Grade Analytical
Packaging Plastic Drum
Density 2.05
Chemical Name or Material Nickel(II) nitrate hexahydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful if swallow
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Avoid release to the e
RTECSNumber QR7300000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.