Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic acid, ACS reagent

Catalog No. p-4440186
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 79-09-4 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Melting Point -22.0°C
Density 0.9900g/mL
Boiling Point 141.0°C
Flash Point 51°C
Packaging Glass bottle
Refractive Index 1.3850 to 1.3870
Linear Formula C2H5CO2H
Quantity 100 mL
Merck Index 15,7939
Specific Gravity 0.99
Solubility Information Solubility in water: soluble.
Viscosity 1.02 mPa.s (25°C)
Formula Weight 74.08
Grade ACS Reagent
Chemical Name or Material Propionic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 201-176-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.