Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Propionic Acid, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mL
5 mL
Description
Propionic acid (PA) is a naturally occurring carboxylic acid, which in its pure state exists as a colorless corrosive liquid with an unpleasant odor. It is miscible in water. Industrially it is produced by hydrocarboxylation of ethylene in presence nickel carbonyl as a catalyst. PA has been found to reduce food intake, lower the fatty acids content in plasma and liver, might improve tissue insulin sensitivity and exerts immunosuppressive actions. Propionic acid is an excellent raw material as it is stable, cheap and safe and can even be used as a food additive.

Specifications
Specifications
CAS | 79-09-4 |
Vapor Pressure | 2.4 mmHg (20°C) |
Vapor Density | 2.55 (vs. air) |
Density | 0.993 g/mL (at 25°C) |
Physical Form | Neat |
Linear Formula | CH3CH2COOH |
MDL Number | MFCD00002756 |
Melting Point | -24°C to +23°C |
Boiling Point | 141°C |
Synonym | Acid C3; Propanoic acid; Propanyl acid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction