Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionic Acid, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

79-09-4
C3H6O2
74.079
XBDQKXXYIPTUBI-UHFFFAOYSA-N
1032
CHEBI:30768
propanoic acid
CCC(=O)O

A grade of chemicals for laboratory use

Packaging Narrow Mouth Amber Glass Bottle
Formula Weight 74.08
Flash Point 52.00°C
Quantity 500mL
Specific Gravity 1L = 0.99kg
Chemical Name or Material Propionic Acid