Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionitrile, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
107-12-0
C3H5N
55.08
MFCD00001948
FVSKHRXBFJPNKK-UHFFFAOYSA-N
propionitrile,ethyl cyanide,propiononitrile,cyanoethane,propylnitrile,propionic nitrile,ether cyanatus,hydrocyanic ether,ethylkyanid,propannitril
7854
CHEBI:26307
propanenitrile
CCC#N
107-12-0
C3H5N
55.08
MFCD00001948
FVSKHRXBFJPNKK-UHFFFAOYSA-N
propionitrile,ethyl cyanide,propiononitrile,cyanoethane,propylnitrile,propionic nitrile,ether cyanatus,hydrocyanic ether,ethylkyanid,propannitril
7854
CHEBI:26307
propanenitrile
CCC#N
Viscosity 0.44 mPa.s (20°C)
Specific Gravity 0.77
Packaging Glass bottle
Density 0.7700g/mL
Melting Point -93.0°C
Quantity 100mL
Flash Point 6°C
Boiling Point 97.0°C
Refractive Index 1.3650 to 1.3670
Assay Percent Range 98.5% min. (GC)
Linear Formula C2H5CN
Beilstein 02,245
Merck Index 15,7941
Solubility Information Solubility in water: 99.4g/L (20°C). Other solubilities: miscible with alcohol, ether, dmf, acetone,, benzene and dichloroethylen
Formula Weight 55.08
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Propionitrile
Show More Show Less

GHS H Statement
Toxic if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:

GHS Signal Word: Danger

EINECSNumber : 203-464-4