Learn More
Propionyl chloride, 98%
Description
Propionyl chloride is used as an intermediate in the production of agrochemicals and pharmaceuticals. It serves as an intermediate for dyes, textile auxiliaries and peroxide compounds. It acts as a crop protecting agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Propionyl chloride is used as an intermediate in the production of agrochemicals and pharmaceuticals. It serves as an intermediate for dyes, textile auxiliaries and peroxide compounds. It acts as a crop protecting agent.
Notes
Moisture sensitive. Incompatible with alcohols, oxidizing agents, strong bases and water.

Specifications
Specifications
Melting Point | -94°C |
Density | 1.061 |
Boiling Point | 77°C to 79°C |
Flash Point | 11°C (51°F) |
Odor | Unpleasant |
Refractive Index | 1.404 |
Linear Formula | CH3CH2COCl |
Quantity | 2500 g |
UN Number | UN1815 |
Beilstein | 385632 |
Show More |
Safety and Handling
Hazard Category | H225-H302-H314-H331-H335 |
Hazard Statement | GHS H Statement H225-H314 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPIONYL CHLORIDE |
EINECSNumber | 201-170-0 |
RTECSNumber | UC3926000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.