Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionyl chloride, 98%

Catalog No. AC131530025
Click to view available options
Quantity:
25 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle

Catalog No. AC131530025

Supplier: Thermo Scientific Chemicals 131530025

Only null left
Add to Cart

CAS: 79-03-8 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-03-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
InChI Key RZWZRACFZGVKFM-UHFFFAOYSA-N
Synonym propionyl chloride, propionic acid chloride, propionic chloride, propionylchloride, unii-mb6vl5omb9, mb6vl5omb9, propanoylchloride, propionyl cloride, propanoic chloride, propionyl-chloride
PubChem CID 62324
IUPAC Name propanoyl chloride
SMILES CCC(=O)Cl

Specifications

Melting Point -94.0°C
Density 1.0600g/mL
Boiling Point 77.0°C to 79.0°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4020 to 1.4050
Linear Formula C2H5COCl
Quantity 2.5 kg
Beilstein 02,243
Specific Gravity 1.06
Merck Index 15,7942
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol
Viscosity 0.48 mPa.s (20°C)
Formula Weight 92.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Propionyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Highly flammable liquid and vapor.
Harmful if swallowed.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 201-170-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.