Learn More
Description
- Polymer case with rubber face cap and unbreakable polycarbonate lens is waterproof and shock and corrosion resistant
- One-handed push-button operation
- Dual switch for constant-stream or momentary-blink operation
- Xenon bulb
2AA ProPolymer:
Operates up to four hours on two AA alkaline batteries (included). Length: 6 in. (15.2cm). Weight: 3.7 oz. (105g). Color: Yellow.
3C ProPolymer:
Operates 5.5 hours on three C alkaline batteries (not included). Length: 8.5 in. (21.6cm). Weight: 13.5 oz. (383g). Color: Yellow.
4AA ProPolymer:
Operates up to four hours on four AA alkaline batteries (included). Length: 6.5 in. (16.5cm). Weight: 6.3 oz. (179g). Color: Yellow.
Order Info
Black fire helmet clamps available for models 2AA and 4AA.
Certifications
UL listed for certain Class I, Division 1 hazardous locations.
Specifications
Specifications
| Gas Fill | Xenon |
| Weight (English) | 6.3 oz. |
| Weight (Metric) | 0.17 kg |
| Battery Type | 4 AA Alkaline |
| Certifications/Compliance | UL Class I, Division 1 |
| Color | Yellow |
| Controller Type | On/Off |
| For Use With (Application) | For constant-on or blink |
| Includes | 4 AA batteries |
| Length (English) | 6.5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.