Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propper Seracult™ Plus Single Slide Fecal Occult Blood Test
SDP

Catalog No. 1255428A Shop All Propper Manufacturing Co. Products
Change view
Click to view available options
Quantity:
100 Pk.
1000 Pk.
20 Pk.
Type:
Developer
Single Slide
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
12-554-28A 100 Pk. Single Slide
23-246930 20 Pk. Developer
12-554-28B 1000 Pk. Single Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 12-554-28A Supplier Propper Supplier No. 37400100
This item is not returnable. View return policy

Seracult™ Plus Single Slide Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer for use with your most high risk patients. Guaiac Fecal Occult Blood Test to aid in the detection of colorectal cancer. Long sample stability. Fast qualitative results.

  • Seracult™ Plus Fecal Occult Blood Test is a highly sensitive qualitative aid in the diagnosis of colorectal cancer.
  • Designed for routine use in physical examinations in doctors offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
  • Seracult™ Plus was designed in mind for patients who are at an increased risk of disease.
  • Seracult™ Plus Single Slides are ideal for testing inpatient or single samples at home.
  • Seracult™ Plus is available in a variety of configurations to meet all of your needs.
  • Results are ready instantly.
  • Seracult guaiac paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
  • Mailing kits of in-office slides.
  • 28-day sample stability.
  • CLIA Waived.

Specifications

Detectable Analytes Hemoglobin
Sample Type Stool
Sensitivity 0.32 mg/100 mL
Type Single Slide
For Use With (Application) Fecal Occult Blood Detection
Includes 100 Single Slides, 2 x 15 mL Bottles of Developer, 100 Applicators
Quantity 100 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.