Learn More
Propper Seracult™ Plus Single Slide Fecal Occult Blood Test
Shop All Propper Manufacturing Co. Products

Description
- Seracult™ Plus Fecal Occult Blood Test is a highly sensitive qualitative aid in the diagnosis of colorectal cancer.
- Designed for routine use in physical examinations in doctors offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
- Seracult™ Plus was designed in mind for patients who are at an increased risk of disease.
- Seracult™ Plus Single Slides are ideal for testing inpatient or single samples at home.
- Seracult™ Plus is available in a variety of configurations to meet all of your needs.
- Results are ready instantly.
- Seracult guaiac paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
- Mailing kits of in-office slides.
- 28-day sample stability.
- CLIA Waived.
Specifications
Specifications
| Detectable Analytes | Hemoglobin |
| Sample Type | Stool |
| Sensitivity | 0.32 mg/100 mL |
| Type | Single Slide |
| For Use With (Application) | Fecal Occult Blood Detection |
| Includes | 1000 Single Slides, 20 x 15 mL Bottles of Developer, 1000 Applicators |
| Quantity | 1000 Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.