Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propper Seracult™ Fecal Occult Blood Testing Triple Slides for Serial Testing
SDP

Catalog No. 1255426A
Click to view available options
Quantity:
34/Cs.
80/Cs.
Type:
Mailing Kit
Seracult Triple Slide
This item is not returnable. View return policy
This item is not returnable. View return policy

Seracult™ Guaiac Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer. Single or triple slide. Offers long sample stability and fast qualitative results.

  • Seracult™ Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer.
  • Designed for routine use in physical examinations in doctors' offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
  • Seracult™ Triple Slides made consecutive day testing easy and convenient for patients.
  • Seracult™ is available in a variety of configurations to meet all of your needs.
  • Results are ready instantly.
  • Seracult paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
  • Detects hemoglobin and products of its degradation.
  • Mailing kits of in-office slides.
  • 28-day sample stability.
  • CLIA waived.

Specifications

Clia Complexity Waived
Detectable Analytes Hemoglobin
Format Kit, In vitro Diagnostic Test
Sample Type Stool
Type Seracult Triple Slide
For Use With (Application) Fecal Occult Blood Detection
Includes 34 Triples Slides, 2 x 15 mL Bottles of Developer, 102 Applicators
Quantity 34/Cs.

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.