Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propranolol Hydrochloride, USP, 98-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18605889
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

PR140, 318-98-9, C16H21NO2·HCl

Spectrum™ Chemical Propranolol Hydrochloride, USP is a sympatholytic nonselective beta blocker used to treat anxiety, hypertension and panic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 318-98-9
Molecular Formula C16H22ClNO2
Molecular Weight (g/mol) 295.81
InChI Key ZMRUPTIKESYGQW-UHFFFAOYNA-N
IUPAC Name hydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
SMILES [H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 162-165°C
Packaging Amber Glass Bottle
Quantity 5 g
Specific Rotation -1.0° to +1.0°
Percent Purity 98 to 101.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.