Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proprotein Convertase 1/PCSK1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298925JF646
Description
Proprotein Convertase 1/PCSK1 Polyclonal antibody specifically detects Proprotein Convertase 1/PCSK1 in Human samples. It is validated for ELISASpecifications
Proprotein Convertase 1/PCSK1 | |
Polyclonal | |
ELISA | |
BMIQ12;NEC1;Neuroendocrine convertase 1;PC1;PC1/3;PC3;SPC3 | |
Produced in rabbits immunized with purified, recombinant Human Proprotein Convertase 1/PCSK1 (Accession#: NP_000430.3; Met 1-Arg 617) | |
0.1 mL | |
Golgi Apparatus Markers | |
5122 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction