Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proprotein Convertase 2/PCSK2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Proprotein Convertase 2/PCSK2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP169146
SDP
View Documents
Novus Biologicals
NBP169146
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Proprotein Convertase 2/PCSK2 Polyclonal specifically detects Proprotein Convertase 2/PCSK2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Proprotein Convertase 2/PCSK2
Polyclonal
Rabbit
Chromatin Research, Golgi Apparatus Markers, Neuroscience
5126
Synthetic peptides corresponding to PCSK2 (proprotein convertase subtilisin/kexin type 2) The peptide sequence was selected from the middle region of PCSK2. Peptide sequence LASTFSNGRKRNPEAGVATTDLYGNCTLRHSGTSAAAPEAAGVFALALEA.
Primary
Western Blot
Unconjugated
RUO
EC 3.4.21, EC 3.4.21.94, KEX2-like endoprotease 2, NEC 2, NEC2SPC2, neuroendocrine convertase 2, PC2NEC-2, Prohormone convertase 2, Proprotein convertase 2, proprotein convertase subtilisin/kexin type 2
PCSK2
IgG
58 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.