Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proprotein Convertase 7/PCSK7 Antibody (OTI1B8), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273621AF700
Description
Proprotein Convertase 7/PCSK7 Monoclonal specifically detects Proprotein Convertase 7/PCSK7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Proprotein Convertase 7/PCSK7 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PCSK7 | |
Full length recombinant protein of human Proprotein Convertase 7/PCSK7 (NP_004707) produced in HEK293T cells. | |
0.1 mL | |
9159.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1B8 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.61, EC 3.4.21.75, EC 3.4.21.93, hPC8, LPCFLJ23503, Lymphoma proprotein convertase, PC7FLJ45329, PC8prohormone convertase PC7, Prohormone convertase 7, Proprotein convertase 7, Proprotein convertase 8, proprotein convertase subtilisin/kexin type 7, SPC7proprotein convertase PC7, Subtilisin/kexin-like protease PC7 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction