Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proprotein Convertase 9/PCSK9 Antibody (007), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290833AF647
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Rat samples. It is validated for Sandwich ELISA Detection.Specifications
Proprotein Convertase 9/PCSK9 | |
Monoclonal | |
Alexa Fluor 647 | |
EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Proprotein Convertase 9/PCSK9 (NP_954862.2; Met1-Gln691). | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism | |
255738 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction